2,6-Dichloronicotinonitrile ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 2,6-Dichloropyridine-3-carbonitrile, 2,6-Dichloro-3-cyanopyridine

D4650-200MG D4650-1G
TCIAD4650-200MGEA 230 NOK
TCIAD4650-200MG TCIAD4650-1G
2,6-Dichloronicotinonitrile ≥98.0% (by GC)
2,6-Dichloronicotinonitrile
Formel: C₆H₂Cl₂N₂
MW: 173,00 g/mol
MDL nummer: MFCD03411341
CAS-nummer: 40381-90-6
FN: 3439
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR