2,6-Dichloro-3-formylpyridine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2,6-Dichloronicotinaldehyde, 2,6-Dichloropyridine-3-carboxaldehyde, 2,6-Dichloro-3-formylpyridine, 2,6-Dichloro-3-pyridinecarboxaldehyde
D4651-1G
D4651-5G
TCIAD4651-1GEA
485
NOK
TCIAD4651-1G
TCIAD4651-5G
2,6-Dichloro-3-formylpyridine ≥98.0% (by GC)
2,6-Dichloro-3-formylpyridine
Formel:
C₆H₃Cl₂NO MW: 176,0001 g/mol Kokepunkt: 95°C |
MDL nummer:
MFCD07369746 CAS-nummer: 55304-73-9 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...