3,6-Dibromo-2,7-dihydroxynaphthalene ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 3,6-Dibromonaphthalene-2,7-diol

D4669-1G D4669-5G
TCIAD4669-1GEA 1550 NOK
TCIAD4669-1G TCIAD4669-5G
3,6-Dibromo-2,7-dihydroxynaphthalene ≥98.0% (by GC)
3,6-Dibromo-2,7-dihydroxynaphthalene
Formel: C₁₀H₆Br₂O₂
MW: 317,98 g/mol
MDL nummer: MFCD00277643
CAS-nummer: 96965-79-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 186.0 to 190.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR