1,4-Diphenyl-2-butyne-1,4-dione ≥96.0% (by GC)

Leverandør: TCI

D4699-250MG D4699-1G
TCIAD4699-250MGEA 945 NOK
TCIAD4699-250MG TCIAD4699-1G
1,4-Diphenyl-2-butyne-1,4-dione ≥96.0% (by GC)
1,4-Diphenyl-2-butyne-1,4-dione
Formel: C₁₆H₁₀O₂
Lagringstemperatur: Kjøleskap
CAS-nummer: 1087-09-8

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 111.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR