α-Chloro-2,5-difluorotoulene ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
D4726-5G
D4726-25G
TCIAD4726-5GEA
610
NOK
TCIAD4726-5G
TCIAD4726-25G
α-Chloro-2,5-difluorotoulene ≥98.0% (by GC)
α-Chloro-2,5-difluorotoulene
Formel:
C₇H₅ClF₂ MW: 162,57 g/mol Kokepunkt: 74°C |
MDL nummer:
MFCD01090989 CAS-nummer: 495-07-8 FN: 3265 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...