dibenzo[b,d]furan-2-carbaldehyde ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
dibenzo[b,d]furan-2-carbaldehyde, Dibenzofuran-2-carboxaldehyde
D4759-1G
D4759-5G
TCIAD4759-1GEA
1225
NOK
TCIAD4759-1G
TCIAD4759-5G
dibenzo[b,d]furan-2-carbaldehyde ≥98.0% (by GC)
dibenzo[b,d]furan-2-carbaldehyde
Formel:
C₁₃H₈O₂ MW: 196,205 g/mol |
MDL nummer:
MFCD00136272 CAS-nummer: 5397-82-0 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...