dibenzo[b,d]thiophene-4-carbaldehyde ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Dibenzothiophene-4-carboxaldehyde, Dibenzo[b,d]thiophene-4-carbaldehyde, Dibenzothiophene-4-carbaldehyde
D4767-1G
D4767-5G
TCIAD4767-1GEA
1050
NOK
TCIAD4767-1G
TCIAD4767-5G
dibenzo[b,d]thiophene-4-carbaldehyde ≥98.0% (by GC)
dibenzo[b,d]thiophene-4-carbaldehyde
Formel:
C₁₃H₈OS MW: 212,266 g/mol |
MDL nummer:
MFCD00961967 CAS-nummer: 23985-81-1 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 125.0 to 129.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...