3',4'-Dihydroxyflavone ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Synonymer:
2-(3,4-Dihydroxyphenyl)-4-benzopyrone, 2-(3,4-Dihydroxyphenyl)chromen-4-one
D4884-200MG
D4884-1G
TCIAD4884-200MGEA
1175
NOK
TCIAD4884-200MG
TCIAD4884-1G
3',4'-Dihydroxyflavone ≥97.0% (by titrimetric analysis)
3',4'-Dihydroxyflavone
Formel:
C₁₅H₁₀O₄ MW: 254,24 g/mol |
MDL nummer:
MFCD00017600 CAS-nummer: 4143-64-0 |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 246.0 to 250.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...