4',5'-Dichloro-2'-nitroacetanilide ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: N-(4,5-Dichloro-2-nitrophenyl)acetamide

D4907-5G D4907-25G
TCIAD4907-5GEA 905 NOK
TCIAD4907-5G TCIAD4907-25G
4',5'-Dichloro-2'-nitroacetanilide ≥97.0% (by GC)
4',5'-Dichloro-2'-nitroacetanilide
Formel: C₈H₆Cl₂N₂O₃
MW: 249,0508 g/mol
MDL nummer: MFCD00024301
CAS-nummer: 5462-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3aee37d98fb3890fbde90c9e15fac497f17ca415&vid=3aee37d98fb3890fbde90c9e15fac497f17ca415&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 97.0 %
Melting point 124.0 to 128.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR