3,4-Dichlorophenyl isothiocyanate ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
1,2-Dichloro-4-isothiocyanatobenzene, 3,4-Dichlorophenyl Isothiocyanate
D4926-5G
D4926-25G
TCIAD4926-5GEA
580
NOK
TCIAD4926-5G
TCIAD4926-25G
3,4-Dichlorophenyl isothiocyanate ≥98.0% (by GC)
3,4-Dichlorophenyl isothiocyanate
Formel:
C₇H₃Cl₂NS MW: 204,08 g/mol Kokepunkt: 148°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00037828 CAS-nummer: 6590-94-9 EINECS: 229-526-0 FN: 1760 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...