2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: 4,9-Dibromo-2,7-bis(2-ethylhexyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone

D4945-1G D4945-5G
TCIAD4945-1GEA 1550 NOK
TCIAD4945-1G TCIAD4945-5G
2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide ≥98.0% (by HPLC, total nitrogen)
2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide
Formel: C₃₀H₃₆Br₂N₂O₄
MW: 648,44 g/mol
MDL nummer: MFCD27923032
CAS-nummer: 1088205-02-0

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 243.0 to 247.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR