4,4-Diphenyl-2-cyclohexen-1-one ≥98.0% (by GC)

Leverandør: TCI

D4982-1G D4982-5G
TCIAD4982-1GEA 1300 NOK
TCIAD4982-1G TCIAD4982-5G
4,4-Diphenyl-2-cyclohexen-1-one ≥98.0% (by GC)
4,4-Diphenyl-2-cyclohexen-1-one
Formel: C₁₈H₁₆O
MDL nummer: MFCD00075159
CAS-nummer: 4528-64-7

Bestill nå

Spesifikasjon Testresultater

Purity (GC) min. 98.0 %
Melting point 94.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR