2,6-Dibromo-9,10-diphenylanthracene ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

D4986-1G D4986-200MG
TCIAD4986-1GEA 5050 NOK
TCIAD4986-1G TCIAD4986-200MG
2,6-Dibromo-9,10-diphenylanthracene ≥98.0% (by HPLC, titration analysis)
2,6-Dibromo-9,10-diphenylanthracene
Formel: C₂₆H₁₆Br₂
MW: 488,22 g/mol
CAS-nummer: 528609-98-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR