2,6-Difluoro-3-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
|
Warning
|
D5006-1G
D5006-5G
TCIAD5006-1GEA
555
NOK
TCIAD5006-1G
TCIAD5006-5G
2,6-Difluoro-3-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2,6-Difluoro-3-methoxyphenylboronic acid
Formel:
C₇H₇BF₂O₃ MW: 187,94 g/mol |
MDL nummer:
MFCD06657880 CAS-nummer: 870779-02-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...