2,3-Difluoro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
|
Warning
|
Synonymer:
2,3-Difluoro-4-methoxybenzeneboronic acid, (2,3-Difluoro-4-methoxyphenyl)boronic acid
D5016-5G
D5016-25G
TCIAD5016-5GEA
635
NOK
TCIAD5016-5G
TCIAD5016-25G
2,3-Difluoro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2,3-Difluoro-4-methoxyphenylboronic acid
Formel:
C₇H₇BF₂O₃ MW: 187,94 g/mol |
MDL nummer:
MFCD07368234 CAS-nummer: 170981-41-6 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...