Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine ≥98.0% (by HPLC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Dicyclohexylphosphino-2’,6’-dimethoxybiphenyl, SPhos, Dicyclohexyl(2',6'-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl, 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl
D5036-1G
D5036-5G
TCIAD5036-1GEA
525
NOK
TCIAD5036-1G
TCIAD5036-5G
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine ≥98.0% (by HPLC)
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine
Formel:
C₂₆H₃₅O₂P MW: 410,53 g/mol |
MDL nummer:
MFCD05861611 CAS-nummer: 657408-07-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 163.0 to 167.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...