2,7-Dibromophenanthrene ≥98.0% (by GC)

Leverandør: TCI

D5050-200MG D5050-1G
TCIAD5050-200MGEA 1375 NOK
TCIAD5050-200MG TCIAD5050-1G
2,7-Dibromophenanthrene ≥98.0% (by GC)
2,7-Dibromophenanthrene
Formel: C₁₄H₈Br₂
MW: 336,03 g/mol
MDL nummer: MFCD09909861
CAS-nummer: 62325-30-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 203.0 to 207.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR