O-(2,4-Dinitrophenyl)hydroxylamine ≥97.0% (by GC)

Leverandør: TCI
Danger

D5103-1G D5103-5G
TCIAD5103-1GEA 380 NOK
TCIAD5103-1G TCIAD5103-5G
O-(2,4-Dinitrophenyl)hydroxylamine ≥97.0% (by GC)
O-(2,4-Dinitrophenyl)hydroxylamine
Formel: C₆H₅N₃O₅
MW: 199,12 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00075001
CAS-nummer: 17508-17-7
EINECS: 241-512-6
FN: 3224
ADR: 4.1,

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR