2,2,3,3,4,4,5,5,6,6-Decafluoro-6-[(1,2,2-trifluorovinyl)oxy]hexanenitrile ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
D5223-5G
D5223-25G
TCIAD5223-5GEA
2125
NOK
TCIAD5223-5G
TCIAD5223-25G
2,2,3,3,4,4,5,5,6,6-Decafluoro-6-[(1,2,2-trifluorovinyl)oxy]hexanenitrile ≥98.0% (by GC)
2,2,3,3,4,4,5,5,6,6-Decafluoro-6-[(1,2,2-trifluorovinyl)oxy]hexanenitrile
Formel:
C₈F₁₃NO MW: 373,07 g/mol Kokepunkt: 110°C Lagringstemperatur: Kjøleskap |
CAS-nummer:
120903-40-4 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...