2-Decylthiophene ≥97.0% (by GC)

Leverandør: TCI

D5240-1ML D5240-5ML
TCIAD5240-1MLEA 865 NOK
TCIAD5240-1ML TCIAD5240-5ML
2-Decylthiophene ≥97.0% (by GC)
2-Decylthiophene
Formel: C₁₄H₂₄S
MW: 224,41 g/mol
Kokepunkt: 167°C
MDL nummer: MFCD00715761
CAS-nummer: 24769-39-9

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR