2,6-Diamino-N2-(2-aminoethyl)-5-nitropyridine ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 2,6-Diamino-N6-(2-aminoethyl)-3-nitropyridine, N2-(2-Aminoethyl)-5-nitropyridine-2,6-diamine

D5245-50MG D5245-250MG
TCIAD5245-50MGEA 2550 NOK
TCIAD5245-50MG TCIAD5245-250MG
2,6-Diamino-N2-(2-aminoethyl)-5-nitropyridine ≥98.0% (by HPLC)
2,6-Diamino-N2-(2-aminoethyl)-5-nitropyridine
Formel: C₇H₁₁N₅O₂
MW: 197,20 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD18379166
CAS-nummer: 252944-01-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=202cae95772b779a3e8288ce9eff162502187c95&vid=202cae95772b779a3e8288ce9eff162502187c95&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR