1,4-Dibromo-3,5-difluorobenzene ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2,5-Dibromo-1,3-difluorobenzene
D5261-1G
D5261-5G
TCIAD5261-1GEA
785
NOK
TCIAD5261-1G
TCIAD5261-5G
1,4-Dibromo-3,5-difluorobenzene ≥98.0% (by GC)
1,4-Dibromo-3,5-difluorobenzene
Formel:
C₆H₂Br₂F₂ MW: 271,8849 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD07783665 CAS-nummer: 128259-71-2 |
|
Spesifikasjon Testresultater
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
![](/m-no.vwr.com/no_NO/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...