3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane ≥96.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Methyltris(dimethylsiloxy)silane
D5263-5ML
D5263-25ML
TCIAD5263-5MLEA
610
NOK
TCIAD5263-5ML
TCIAD5263-25ML
3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane ≥96.0% (by GC)
3-[(Dimethylsilyl)oxy]-1,1,3,5,5-pentamethyltrisiloxane
Formel:
C₇H₂₄O₃Si₄ MW: 268,61 g/mol Kokepunkt: 164°C |
MDL nummer:
MFCD00054789 CAS-nummer: 17082-46-1 EINECS: 241-136-2 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...