4,6-Dichloronicotinic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4,6-dichloropyridine-3-carboxylic acid
D5311-1G
D5311-5G
TCIAD5311-1GEA
285
NOK
TCIAD5311-1G
TCIAD5311-5G
4,6-Dichloronicotinic acid ≥98.0% (by titrimetric analysis)
4,6-Dichloronicotinic acid
Formel:
C₆H₃Cl₂NO₂ MW: 192,00 g/mol |
MDL nummer:
MFCD00234146 CAS-nummer: 73027-79-9 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...