4,4'-Dibromo-4''-tert-butyltriphenylamine ≥95.0% (by HPLC, total nitrogen)

Leverandør: TCI

Synonymer: N,N-Bis(4-bromophenyl)-4-tert-butylaniline

D5330-1G D5330-5G
TCIAD5330-1GEA 1450 NOK
TCIAD5330-1G TCIAD5330-5G
4,4'-Dibromo-4''-tert-butyltriphenylamine ≥95.0% (by HPLC, total nitrogen)
4,4'-Dibromo-4''-tert-butyltriphenylamine
Formel: C₂₂H₂₁Br₂N
MW: 459,23 g/mol
CAS-nummer: 852534-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c91b04430d273dcabb750cbacca81d2adbef1c8&vid=4c91b04430d273dcabb750cbacca81d2adbef1c8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR