3,6-Dibromo-9-(4-methoxyphenyl)-9H-carbazole ≥98.0% (by HPLC)

Leverandør: TCI
Warning

D5340-1G D5340-5G
TCIAD5340-1GEA 2000 NOK
TCIAD5340-1G TCIAD5340-5G
3,6-Dibromo-9-(4-methoxyphenyl)-9H-carbazole ≥98.0% (by HPLC)
3,6-Dibromo-9-(4-methoxyphenyl)-9H-carbazole
Formel: C₁₉H₁₃Br₂NO
MW: 431,13 g/mol
MDL nummer: MFCD30721572
CAS-nummer: 746651-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5c1daec98e195d1eac0815ac34aa21f2e0f3e2a&vid=c5c1daec98e195d1eac0815ac34aa21f2e0f3e2a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR