Di-tert-butylcarbodiimide ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,3-Di-tert-butylcarbodiimide, N,N'-Di-tert-butylcarbodiimide, N,N-di-tert-butylcarbodiimide, N,N'-methanetetraylbis(1,1-dimethylethylamine)

D5347-5ML D5347-25ML
TCIAD5347-5MLEA 540 NOK
TCIAD5347-5ML TCIAD5347-25ML
Di-tert-butylcarbodiimide ≥98.0% (by GC)
Di-tert-butylcarbodiimide
Formel: C₉H₁₈N₂
MW: 154,26 g/mol
Kokepunkt: 50°C
Flash punkt: 35°C
MDL nummer: MFCD00010231
CAS-nummer: 691-24-7
EINECS: 211-719-6
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac68e1b98b135ce5d701322802e6ae67f088f0e1&vid=ac68e1b98b135ce5d701322802e6ae67f088f0e1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR