1,3-Dimethylimidazolium methyl sulphate ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
|
Warning
|
Synonymer:
1,3-Dimethyl-1H-imidazol-3-ium methyl sulphate
D5348-5G
D5348-25G
TCIAD5348-5GEA
525
NOK
TCIAD5348-5G
TCIAD5348-25G
1,3-Dimethylimidazolium methyl sulphate ≥98.0% (by HPLC, total nitrogen)
1,3-Dimethylimidazolium methyl sulphate
Formel:
C₆H₁₂N₂O₄S MW: 208,24 g/mol |
MDL nummer:
MFCD03453276 CAS-nummer: 97345-90-9 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...