3,5-Dimethoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: 2-(3,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 3,5-Dimethoxyphenylboronic acid Pinacol ester

D5360-1G D5360-5G
TCIAD5360-1GEA 660 NOK
TCIAD5360-1G TCIAD5360-5G
3,5-Dimethoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3,5-Dimethoxyphenylboronic acid pinacol ester
Formel: C₁₄H₂₁BO₄
MW: 264,13 g/mol
MDL nummer: MFCD05865191
CAS-nummer: 365564-07-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77e488b212d40809d4e119553b06236e551f2d20&vid=77e488b212d40809d4e119553b06236e551f2d20&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 88.0 to 92.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR