(S)-2,4-Diphenyl-4,5-dihydrooxazole ≥98.0% (by GC)

Leverandør: TCI

D5368-5G D5368-1G
TCIAD5368-5GEA 4575 NOK
TCIAD5368-5G TCIAD5368-1G
(S)-2,4-Diphenyl-4,5-dihydrooxazole ≥98.0% (by GC)
(S)-2,4-Diphenyl-4,5-dihydrooxazole
Formel: C₁₅H₁₃NO
CAS-nummer: 475571-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbae8cfc31a0cedbe52d28d9b06213a23ed6842e&vid=dbae8cfc31a0cedbe52d28d9b06213a23ed6842e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR