Dorsomorphin ≥97.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 6-(4-(2-(Piperidin-1-yl)ethoxy)phenyl)-3-(pyridin-4-yl)pyrazolo[1,5-a]pyrimidine, 6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine

D5394-10MG D5394-50MG
TCIAD5394-10MGEA 1375 NOK
TCIAD5394-10MG TCIAD5394-50MG
Dorsomorphin ≥97.0% (by HPLC)
Dorsomorphin
Formel: C₂₄H₂₅N₅O
MW: 399,5 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD08705402
CAS-nummer: 866405-64-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f30ef90e218b98654ddfd85528189e29c132e74&vid=7f30ef90e218b98654ddfd85528189e29c132e74&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 16.50 to 18.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR