3,6-Dibromo-9-heptyl-9H-carbazole ≥98.0% (by GC)

Leverandør: TCI
Warning

D5404-1G D5404-5G
TCIAD5404-1GEA 1675 NOK
TCIAD5404-1G TCIAD5404-5G
3,6-Dibromo-9-heptyl-9H-carbazole ≥98.0% (by GC)
3,6-Dibromo-9-heptyl-9H-carbazole
Formel: C₁₉H₂₁Br₂N
MW: 423,19 g/mol
MDL nummer: MFCD31618132
CAS-nummer: 1103535-99-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a16f4502fbdcfd2d0f0855933129f03a1c0d77c0&vid=a16f4502fbdcfd2d0f0855933129f03a1c0d77c0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR