Diclazuril ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 2-(4-Chlorophenyl)-2-(2,6-dichloro-4-(3,5-dioxo-4,5-dihydro-1,2,4-triazin-2(3H)-yl)phenyl)acetonitrile, 2,6-Dichloro-alpha-(4-chlorophenyl)-4-[4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)-yl]benzeneacetonitrile

D5457-1G D5457-5G
TCIAD5457-1GEA 2025 NOK
TCIAD5457-1G TCIAD5457-5G
Diclazuril ≥98.0% (by HPLC, titration analysis)
Diclazuril
Formel: C₁₇H₉Cl₃N₄O₂
MW: 407,64 g/mol
MDL nummer: MFCD00867203
CAS-nummer: 101831-37-2
Merck Indeks: 14,03080

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 295.0 to 299.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR