3,6-Dibromopyridazine ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
D5466-1G
D5466-5G
TCIAD5466-1GEA
300
NOK
TCIAD5466-1G
TCIAD5466-5G
3,6-Dibromopyridazine ≥98.0% (by GC)
3,6-Dibromopyridazine
Formel:
C₄H₂Br₂N₂ MW: 237,88 g/mol |
MDL nummer:
MFCD00233947 CAS-nummer: 17973-86-3 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...