DBeQ ≥98.0% (by HPLC)
Leverandør: TCI
Synonymer:
N,N'-Dibenzylquinazoline-2,4-diamine, N2,N4-Dibenzylquinazoline-2,4-diamine
D5476-100MG
D5476-25MG
TCIAD5476-100MGEA
7725
NOK
TCIAD5476-100MG
TCIAD5476-25MG
DBeQ ≥98.0% (by HPLC)
DBeQ
Formel:
C₂₂H₂₀N₄ MW: 340,43 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD03691820 CAS-nummer: 177355-84-9 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 15.50 to 17.00 % |
Melting point | 147.0 to 151.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...