4'-(Di-p-tolylamino)-[1,1'-biphenyl]-4-yl acrylate ≥97.0% (by HPLC, total nitrogen)
Leverandør: TCI
Synonymer:
2-Propenoic acid 4'-[Bis(4-methylphenyl)amino]-[1,1'-biphenyl]-4-yl ester, 4'-[Bis(4-methylphenyl)amino]-[1,1'-biphenyl]-4-yl 2-Propenoate, Acrylic acid 4'-(Di-p-tolylamino)-[1,1'-biphenyl]-4-yl ester
D5507-1G
D5507-5G
TCIAD5507-1GEA
700
NOK
TCIAD5507-1G
TCIAD5507-5G
4'-(Di-p-tolylamino)-[1,1'-biphenyl]-4-yl acrylate ≥97.0% (by HPLC, total nitrogen)
4'-(Di-p-tolylamino)-[1,1'-biphenyl]-4-yl acrylate
Formel:
C₂₉H₂₅NO₂ MW: 419,52 g/mol |
MDL nummer:
MFCD28975082 CAS-nummer: 152636-45-8 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...