Dibenzo[b,d]furan-4-amine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Dibenzofuranamine, 4-Aminodibenzofuran
D5520-1G
D5520-5G
TCIAD5520-1GEA
1100
NOK
TCIAD5520-1G
TCIAD5520-5G
Dibenzo[b,d]furan-4-amine ≥98.0% (by GC, titration analysis)
Dibenzo[b,d]furan-4-amine
Formel:
C₁₂H₉NO MW: 183,21 g/mol Smeltepunkt: 85°C |
MDL nummer:
MFCD11110988 CAS-nummer: 50548-43-1 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...