3,4-Dimethoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Leverandør: TCI

Synonymer: 3,4-Dimethoxyphenylboronic acid Pinacol ester, 1,2-Dimethoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-(3,4-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

D5533-1G D5533-5G
TCIAD5533-1GEA 420 NOK
TCIAD5533-1G TCIAD5533-5G
3,4-Dimethoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3,4-Dimethoxyphenylboronic acid pinacol ester
Formel: C₁₄H₂₁BO₄
MW: 264,13 g/mol
Smeltepunkt: 83°C
MDL nummer: MFCD05863909
CAS-nummer: 365564-10-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR