Diethyl 2,6-Dimethylpyridine-3,5-dicarboxylate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Diethyl 2,6-dimethylpyridine-3,5-dicarboxylate, 2,6-Dimethylpyridine-3,5-dicarboxylic acid Diethyl ester

D5549-1G D5549-5G
TCIAD5549-1GEA 730 NOK
TCIAD5549-1G TCIAD5549-5G
Diethyl 2,6-Dimethylpyridine-3,5-dicarboxylate ≥98.0% (by GC)
Diethyl 2,6-Dimethylpyridine-3,5-dicarboxylate
Formel: C₁₃H₁₇NO₄
MW: 251,28 g/mol
Kokepunkt: 208°C
MDL nummer: MFCD00023506
CAS-nummer: 1149-24-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR