1,5-Diacetyl-1,3,5-triazinane-2,4-dione ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

D5634-1G D5634-5G
TCIAD5634-1GEA 1125 NOK
TCIAD5634-1G TCIAD5634-5G
1,5-Diacetyl-1,3,5-triazinane-2,4-dione ≥98.0% (by HPLC, titration analysis)
1,5-Diacetyl-1,3,5-triazinane-2,4-dione
Formel: C₇H₉N₃O₄
MW: 199,17 g/mol
MDL nummer: MFCD00604438
CAS-nummer: 86320-44-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 212.0 to 216.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR