Dansylsarcosine Cyclohexylammonium salt ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Synonymer:
Dansylsarcosine Cyclohexylamine salt, Dns-Me-Gly-OH.Cyclohexylammonium, 5-Dimethylaminonaphthalene-1-sulphonylsarcosine Cyclohexylammonium salt, N-(5-Dimethylaminonaphthalene-1-sulphonyl)-N-methylglycine Cyclohexylammonium salt
D5722-50MG
D5722-250MG
TCIAD5722-50MGEA
1100
NOK
TCIAD5722-50MG
TCIAD5722-250MG
Dansylsarcosine Cyclohexylammonium salt ≥98.0% (by titrimetric analysis)
Dansylsarcosine Cyclohexylammonium salt
Formel:
C₁₅H₁₈N₂O₄S·C₆H₁₃N MW: 421,56 g/mol Lagringstemperatur: Fryseskap |
CAS-nummer:
2733745-64-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...