Dipropyl trisulphide ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Dipropyltrisulphane

D5750-25G D5750-5G
TCIAD5750-25GEA 1100 NOK
TCIAD5750-25G TCIAD5750-5G
Dipropyl trisulphide ≥98.0% (by GC)
Dipropyl trisulphide
Formel: C₆H₁₄S₃
MW: 182,36 g/mol
Kokepunkt: 72°C
MDL nummer: MFCD00040074
CAS-nummer: 6028-61-1
EINECS: 227-903-4

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR