3,5-Dichloro-4-pyridinecarboxaldehyde ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3,5-Dichloro-4-formylpyridine, 3,5-Dichloropyridine-4-carboxaldehyde, 3,5-Dichloroisonicotinaldehyde

D5790-1G D5790-5G
TCIAD5790-1GEA 300 NOK
TCIAD5790-1G TCIAD5790-5G
3,5-Dichloro-4-pyridinecarboxaldehyde ≥98.0% (by GC)
3,5-Dichloro-4-pyridinecarboxaldehyde
Formel: C₆H₃Cl₂NO
MW: 176,00 g/mol
MDL nummer: MFCD04039921
CAS-nummer: 136590-83-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR