2,5-Dibromoterephthalaldehyde ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2,5-Dibromobenzene-1,4-dicarbaldehyde, 2,5-Dibromo-1,4-diformylbenzene, 2,5-Dibromo-1,4-benzenedicarboxaldehyde

D5793-1G D5793-250MG
TCIAD5793-1GEA 2250 NOK
TCIAD5793-1G TCIAD5793-250MG
2,5-Dibromoterephthalaldehyde ≥97.0% (by GC)
2,5-Dibromoterephthalaldehyde
Formel: C₈H₄Br₂O₂
MW: 291,93 g/mol
MDL nummer: MFCD11521314
CAS-nummer: 63525-48-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 189.0 to 193.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR