Sodium-4-ethylbenzenesulphonate ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
4-Ethylbenzenesulphonic acid Sodium Salt
E0104-25G
E0104-500G
TCIAE0104-25GEA
700
NOK
TCIAE0104-25G
TCIAE0104-500G
Sodium-4-ethylbenzenesulphonate ≥98.0% (by HPLC, titration analysis)
Sodium-4-ethylbenzenesulphonate
Formel:
C₈H₁₀O₃S MW: 208,21 g/mol |
MDL nummer:
MFCD00070568 CAS-nummer: 14995-38-1 EINECS: 239-080-9 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...