Ethylene glycol monoisobutyl ether ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
Isobutyl Cellosolve, 2-Isobutoxyethanol
E0301-25ML
E0301-500ML
TCIAE0301-25MLEA
230
NOK
TCIAE0301-25ML
TCIAE0301-500ML
Ethylene glycol monoisobutyl ether ≥98.0% (by GC)
Ethylene glycol monoisobutyl ether
Formel:
C₆H₁₄O₂ MW: 118,18 g/mol Kokepunkt: 160°C Flash punkt: 57°C |
MDL nummer:
MFCD00059601 CAS-nummer: 4439-24-1 EINECS: 224-658-5 FN: 1992 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...