Estradiol benzoate ≥97.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
β-Estradiol-3-benzoate
E0329-1G
E0329-5G
TCIAE0329-1GEA
700
NOK
TCIAE0329-1G
TCIAE0329-5G
Estradiol benzoate ≥97.0% (by GC)
Estradiol benzoate
Formel:
C₂₅H₂₈O₃ MW: 376,496 g/mol |
MDL nummer:
MFCD00003692 CAS-nummer: 50-50-0 EINECS: 200-043-7 Merck Indeks: 14,03703 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 190.0 to 196.0 deg-C |
Specific rotation [a]20/D | +58.0 to +63.0 deg (C=1, Dioxane) |
Solubility in Dioxane | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...