3-Ethoxy-2-cyclohexen-1-one ≥98.0% (by GC)

Leverandør: TCI

E0384-25ML E0384-5ML
TCIAE0384-25MLEA 2050 NOK
TCIAE0384-25ML TCIAE0384-5ML
3-Ethoxy-2-cyclohexen-1-one ≥98.0% (by GC)
3-Ethoxy-2-cyclohexen-1-one
Formel: C₈H₁₂O₂
MW: 140,18 g/mol
Flash punkt: 107°C
MDL nummer: MFCD00001580
CAS-nummer: 5323-87-5
EINECS: 226-190-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR