Ethylenedioxybis(trimethylsilane) ≥97.0% (by GC) protecting reagent for aldehydes and ketones

Leverandør: TCI
Warning

Synonymer: 1,2-Bis(trimethylsilyloxy)ethane, Ethylene Glycol Bis(trimethylsilyl Ether), 1,2-Bis(trimethylsiloxy)ethane

E0478-25G E0478-5G
TCIAE0478-25GEA 1050 NOK
TCIAE0478-25G TCIAE0478-5G
Ethylenedioxybis(trimethylsilane) ≥97.0% (by GC) protecting reagent for aldehydes and ketones
Ethylenedioxybis(trimethylsilane)
Formel: C₈H₂₂O₂Si₂
MW: 206,43 g/mol
Kokepunkt: 166°C
Flash punkt: 46°C
MDL nummer: MFCD00009640
CAS-nummer: 7381-30-8
EINECS: 230-950-3
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR