1-Ethylpyrrolidine ≥98.0% (by GC)
Leverandør: TCI
Danger
Synonymer: N-Ethylpyrrolidine
E0510-25G E0510-5G
TCIAE0510-25GEA 2500 NOK
TCIAE0510-25G TCIAE0510-5G
1-Ethylpyrrolidine ≥98.0% (by GC)
1-Ethylpyrrolidine
Formel: C₆H₁₃N
MW: 99,18 g/mol
Kokepunkt: 105°C
MDL nummer: MFCD01632186
CAS-nummer: 7335-06-0
EINECS: 230-840-5
FN: 2733
ADR: 3,II
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %